Learn More
ATCC Acinetobacter baumannii; Strain AYE
Catalog No. 502381381
Encompass
Acinetobacter baumannii strain AYE is a multidrug-resistant bacterium that was isolated in 2003 from human blood. This whole-genome sequenced strain has applications in antimicrobial resistance research.
Catalog No. 50-238-1381 Supplier ATCC Supplier No. BAA1710
Acinetobacter baumannii strain AYE is a multidrug-resistant bacterium that was isolated in 2003 from human blood. This whole-genome sequenced strain has applications in antimicrobial resistance research.