Learn More
ATCC Salmonella enterica; subsp. diarizonae
Supplier:  ATCC 12325
Encompass
Type strain CDC. Formerly Salmonella choleraesuis subsp. diarizonae. Control strain for detection of Salmonella.
Catalog No. 50-238-3141