Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Montevideo
Supplier:  ATCC 8387
Encompass
Salmonella enterica subsp. enterica serovar Montevideo strain 623 is a whole-genome sequenced bacterium with applications in enteric disease and zoonotic disease research. This strain is typed as I 6,7:g,m,s: -.
Catalog No. 50-238-1078
 Restricted Item