Learn More
ATCC Shigella flexneri; 2457T; genomic DNA
Supplier:  ATCC 700930D5
Encompass
Genomic DNA isolated from Shigella flexneri strain 2457T (ATCC 700930). The presence of the plasmid pINV-2457T has not been confirmed.
Catalog No. 50-238-1644