Learn More
ATCC Haemophilus ducreyi; 35000 HP; genomic DNA
Supplier:  ATCC 700724D5
Encompass
Genomic DNA isolated from Haemophilus ducreyi strain 35000 HP. This whole-genome sequenced product can be used in PCR and other molecular biology applications, infectious disease research, and sexually transmitted disease research.
Catalog No. 50-238-1263