Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Typhimurium
Supplier:  ATCC 14028
Encompass
Salmonella enterica subsp. enterica serovar Typhimurium strain CDC 6516-60 is a whole-genome sequenced bacterium that was isolated from the pooled heart and liver tissue of four-week-old chickens. This bacterium has applications in enteric disease research, media testing, and water testing.
Catalog No. 50-238-4083