Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Newport
Catalog No. 502381828
Encompass
Salmonella enterica subsp. enterica serovar Newport strain NCTC 129 was isolated from a food poisoning fatality in England. This whole-genome sequenced bacterial strain is typed as serotype I 6,8:e,h:1,2 and it can be used in enteric disease research, infectious disease research, and zoonotic disease research.
Catalog No. 50-238-1828 Supplier ATCC Supplier No. 6962
 Restricted Item
Add to Cart
Add to Cart
Salmonella enterica subsp. enterica serovar Newport strain NCTC 129 was isolated from a food poisoning fatality in England. This whole-genome sequenced bacterial strain is typed as serotype I 6,8:e,h:1,2 and it can be used in enteric disease research, infectious disease research, and zoonotic disease research.