Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Typhimurium
Supplier:  ATCC 29630
Encompass
Salmonella enterica subsp. enterica strain TA 1537 is a whole-genome sequenced strain with applications in enteric disease research, infectious disease research, and zoonotic disease research. This bacterium is the HisC3076, rfa-, uvrB derivative of strain LT-2.
Catalog No. 50-238-3009