Learn More
ATCC Salmonella enterica; subsp. enterica Newport
Catalog No. 502382969
Encompass
Salmonella enterica subsp. enterica serovar Newport is a whole-genome sequenced strain that was isolated in 2012 from water in Pennsylvania. It has applications in enteric, infectious, and zoonotic disease research.
Catalog No. 50-238-2969 Supplier ATCC Supplier No. BAA3138
Salmonella enterica subsp. enterica serovar Newport is a whole-genome sequenced strain that was isolated in 2012 from water in Pennsylvania. It has applications in enteric, infectious, and zoonotic disease research.