Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Choleraesuis
Supplier:  ATCC 13312
Encompass
Salmonella enterica subsp. enterica serovar Choleraesuis strain NCTC 5735 [1348, K.34] is a whole-genome sequenced bacterium with applications in quality control, enteric disease research, infectious disease research, and zoonotic disease research. This strain is typed as serotype I 6,7:c:1,5.
Catalog No. 50-238-2572