Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Paratyphi A
Supplier:  ATCC 11511
Encompass
Salmonella enterica subsp. enterica serovar Paratyphi A strain CDC has applications in enteric disease research and zoonotic disease research. This bacterium is serotype 2,12:a:-.
Catalog No. 50-238-4391