Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Typhimurium
Supplier:  ATCC 13311
Encompass
Salmonella enterica subsp. enterica serovar Typhimurium strain NCTC 74 is a whole-genome sequenced bacterial strain isolated from feces in a food poisoning case. The culture applications in bioinformatics, enteric disease research, infectious disease research, and media testing.
Catalog No. 50-238-4578