Learn More
ATCC Paenibacillus glucanolyticus
Catalog No. 502379898
Encompass
Paenibacillus glucanolyticus strain S93 is a bacterial type strain that was isolated from soil in England. This culture is cited to degrade beta glucans.
Catalog No. 50-237-9898 Supplier ATCC Supplier No. 49278
Paenibacillus glucanolyticus strain S93 is a bacterial type strain that was isolated from soil in England. This culture is cited to degrade beta glucans.