Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella enterica subsp. enterica serovar Abony derived from NCTC 6017, Epower
SDP

Catalog No. 10810309 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-810-309 10 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-810-309 Supplier Microbiologics™ Supplier No. 0890E7
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epower is the embodiment of flexibility and versatility. Use one product for all of your QC needs – from media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing. Use a single strain or combine multiple strains for a mixed population.

  • Biosafety Level: 2
  • 10 pellets of a single microorganism with a predetermined quantitative assayserovar Abony
  • Mean Assay Value: 1.0-9.9E+07 CFU per pellet

Specifications

ATCC® NCTC 6017
Biosafety Level 2
Content And Storage 2°C to 8°C
Format Epower
Manufacturing Origin United States
Product Type QC Microorganism
Packaging Type Plastic Canister
Quantity 10 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.