Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella enterica subsp. enterica serovar Choleraesuis ATCC 10708
SDP

Catalog No. 23017288 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-017-288 5 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-288 Supplier Microbiologics™ Supplier No. 0902X
Add to Cart
Add to Cart
This item is not returnable. View return policy

KWIK-STIK Plus combines microorganisms that are just two passages from their reference strains in the signature KWIK-STIK format.

  • Each device features a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab - everything needed for growing control cultures included in one device.
  • 5 KWIK-STIK™ units with a microorganism limited to 2 passages from the reference culture

Specifications

ATCC® ATCC 10708
Biosafety Level 2
Content And Storage 2°C to 8°C
Format Kwik-Stik Plus
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Disinfectant Qualification Testing, Microbial Identification
Packaging Type Plastic Canister
Quantity 5 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.