Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella enterica subsp. enterica serovar Choleraesuis derived from ATCC 10708
SDP

Catalog No. 10810340 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
1 KWIK-STIK
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-810-340 1 KWIK-STIK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-810-340 Supplier Microbiologics™ Supplier No. 0902CRM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lab-Elite CRM microorganisms are just one passage from the reference culture and are delivered in our signature KWIK-STIK format. Our Lab-Elite CRM is designed to help those labs meet the requirements of ISO 17025 standards.

  • One self-contained device including a lyophilized microorganism pellet, reservoir of hydrating fluid and inoculating swab (KWIK-STIK™ format)

Specifications

ATCC® ATCC 10708
Biosafety Level 2
Certifications/Compliance AOAC Official Methods of Analysis (OMA), ISO/IEC 17025
Content And Storage 2°C to 8°C
Format Lab Elite CRM
Manufacturing Origin United States
Product Type Lab Elite CRM
For Use With (Application) Disinfectant Qualification Testing, Food and Food Ingredients: Pathogen Testing, Microbial Identification
Packaging Type Plastic Canister
Quantity 1 KWIK-STIK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.