Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella Enterica Subsp. Enterica Serovar Typhimurium (Derived from ATCC™ 14028™)
SDP

Catalog No. 23047406 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ATCC®
23-047-406 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-047-406 Supplier Microbiologics™ Supplier No. 0363 PEC

missing translation for 'validMainframeMsg'

Microorganism pellets deliver a final concentration of 1 x 105 to 1 x 106 CFU per 1 mL, along with 2 mL vials of hydrating fluid.

  • Pellets deliver a final concentration of 1 x 105 to 1 x 106CFU per 1 mL
  • Includes 20 Microorganism Pellets, and 10 x 2 mL Vials of Hydration Fluid

Specifications

ATCC® Yes
Biosafety Level 2
Content And Storage 2°C to 8°C
Format EZ-PEC™
Manufacturing Origin United States
Product Type Salmonella enterica subsp. enterica serovar typhimurium
Target Organism Class Bacteria
Test Type Antimicrobial Effectiveness, Preservation Efficacy Testing, Disinfectant Qualification Testing
For Use With (Application) Clinical
Includes Microorganism Pellets (20), Hydrating Fluid (10 x 2 mL Vials)
Strain Designations Group B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.