Learn More
ATCC Nocardioides simplex
Catalog No. 502380554
Encompass
Nocardioides simplex strain NCTC 4215 is a whole-genome sequenced bacterial type strain that was isolated from rice soil.
Catalog No. 50-238-0554 Supplier ATCC Supplier No. 6946
Nocardioides simplex strain NCTC 4215 is a whole-genome sequenced bacterial type strain that was isolated from rice soil.