Learn More
ATCC Salmonella enterica; subspecies enterica; serovar Enteritidis; Strain: SAP18-0432
Catalog No. 502381869
Encompass
Salmonella enterica subsp. enterica serovar Enteritidis strain SAP18-0432 was isolated in the United States from a human. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in infectious disease research and zoonotic disease research.
Catalog No. 50-238-1869 Supplier ATCC Supplier No. BAA3159
Salmonella enterica subsp. enterica serovar Enteritidis strain SAP18-0432 was isolated in the United States from a human. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in infectious disease research and zoonotic disease research.