Learn More
ATCC Salmonella enterica; subspecies enterica; serovar Typhimurium; Strain: SAP17-7699
Catalog No. 502383996
Encompass
Salmonella enterica subsp. enterica serovar Typhimurium strain SAP17-7699 was isolated from human feces in the United States. It has applications in food testing, enteric disease research, and zoonotic disease research.
Catalog No. 50-238-3996 Supplier ATCC Supplier No. BAA3155
Salmonella enterica subsp. enterica serovar Typhimurium strain SAP17-7699 was isolated from human feces in the United States. It has applications in food testing, enteric disease research, and zoonotic disease research.