Learn More
ATCC Salmonella enterica; subspecies enterica; serovar Enteritidis; Strain: SAP18-H9654
Catalog No. 502384092
Encompass
Salmonella enterica subsp. enterica serovar Enteritidis strain SAP18-H9654 was isolated in the United States from human feces. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in infectious disease research and zoonotic disease research.
Catalog No. 50-238-4092 Supplier ATCC Supplier No. BAA3160
Salmonella enterica subsp. enterica serovar Enteritidis strain SAP18-H9654 was isolated in the United States from human feces. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in infectious disease research and zoonotic disease research.