Learn More
ATCC Salmonella enterica; subspecies enterica; serovar Typhimurium; Strain: SAP18-6199
Catalog No. 502381841
Encompass
Salmonella enterica subsp. enterica serovar Typhimurium strain SAP18-6199 was isolated from a human. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in enteric disease research, infectious disease research, and zoonotic disease research.
Catalog No. 50-238-1841 Supplier ATCC Supplier No. BAA3158
Salmonella enterica subsp. enterica serovar Typhimurium strain SAP18-6199 was isolated from a human. This whole-genome sequenced bacterium was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. This strain has applications in enteric disease research, infectious disease research, and zoonotic disease research.