Learn More
Cederlane SALMONELLA H:6 150TST 3ML ANT
SDP
Catalog No. NC0617963
Encompass
(Product: H:6),(No. of tests: 150),(SSI� SALMONELLA H FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0617963 Supplier Cederlane Supplier No. 40342(SS)
(Product: H:6),(No. of tests: 150),(SSI� SALMONELLA H FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial