Learn More
Cederlane SALMONELLA MULTIPHASE HI
SDP
Catalog No. NC0533575
Encompass
Product: H:i),(No. of tests: 150),(SSI� SALMONELLA H PHASE ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
This item is not returnable. View return policy
Catalog No. NC0533575 Supplier Cederlane Supplier No. 23850(SS)
Product: H:i),(No. of tests: 150),(SSI� SALMONELLA H PHASE ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial