Learn More
Cederlane SALMONELLA MULTIPHASE HM
SDP
Catalog No. NC0533572
Encompass
(Product: H:m),(No. of tests: 150),(SSI� SALMONELLA H FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
This item is not returnable. View return policy
Catalog No. NC0533572 Supplier Cederlane Supplier No. 40332(SS)
(Product: H:m),(No. of tests: 150),(SSI� SALMONELLA H FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial