Learn More
Cederlane SALMONELLA MULTIPHASE O4
SDP
Catalog No. NC0533579
Encompass
(Product: O:4),(Reacts with factor:B),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
This item is not returnable. View return policy
Catalog No. NC0533579 Supplier Cederlane Supplier No. 23839(SS)
(Product: O:4),(Reacts with factor:B),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial