Learn More
Cederlane SALMONELLA O:46
SDP
Catalog No. NC0603001
Encompass
(Product: O:46),(Reacts with factor:D2,D3),(No. of tests: 150),(SSI� SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0603001 Supplier Cederlane Supplier No. 40288(SS)
(Product: O:46),(Reacts with factor:D2,D3),(No. of tests: 150),(SSI� SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial