Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cederlane Salmonella O:55 Antiserum, 3 mL vial, 150 tests, Polyclonal antibody, Slide agglutination test, Rabbit host
SDP

Catalog No. NC0617920
Encompass

This technical summary provides an overview of the Salmonella O:55 Antiserum, a diagnostic reagent essential for the serological identification of *Salmonella* species. This polyclonal antibody is tailored for slide agglutination tests, ensuring specificity for the Salmonella O:55 antigen. Each 3 mL vial is capable of performing 150 tests, making it a reliable solution for clinical and research applications in *Salmonella* serotyping.

  • Target antigen is Salmonella O:55
  • Antigenic factor is 55
  • Test method is slide agglutination
  • Antibody type is polyclonal
  • Host animal is rabbit
  • Format is ready-to-use liquid
  • Volume is 3 mL
  • Tests per vial is 150
  • Packaging is dropper bottle
  • Shelf life is up to five years
  • Specificity tested for known O cross-reactions
  • Antigen preparation involves immunization with boiled *Salmonella* culture

Catalog No. NC0617920 Supplier Cederlane Supplier No. 40253(SS)
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This technical summary provides an overview of the Salmonella O:55 Antiserum, a diagnostic reagent essential for the serological identification of *Salmonella* species. This polyclonal antibody is tailored for slide agglutination tests, ensuring specificity for the Salmonella O:55 antigen. Each 3 mL vial is capable of performing 150 tests, making it a reliable solution for clinical and research applications in *Salmonella* serotyping.

  • Target antigen is Salmonella O:55
  • Antigenic factor is 55
  • Test method is slide agglutination
  • Antibody type is polyclonal
  • Host animal is rabbit
  • Format is ready-to-use liquid
  • Volume is 3 mL
  • Tests per vial is 150
  • Packaging is dropper bottle
  • Shelf life is up to five years
  • Specificity tested for known O cross-reactions
  • Antigen preparation involves immunization with boiled *Salmonella* culture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.