Learn More
Cederlane SALMONELLA O ANTISERA SNGL F7
SDP
Catalog No. NC0486849
Encompass
(Product: O:7),(Reacts with factor:C1 ),(No. of tests: 150),(SSI� SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0486849 Supplier Cederlane Supplier No. 40274(SS)
(Product: O:7),(Reacts with factor:C1 ),(No. of tests: 150),(SSI� SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial