Learn More
Cederlane SALMONELLA O ANTISERA SNGL F9
SDP
Catalog No. NC0486850
Encompass
(Product: O:9),(Reacts with factor:D1, D2, D3),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0486850 Supplier Cederlane Supplier No. 40276(SS)
(Product: O:9),(Reacts with factor:D1, D2, D3),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial