Learn More
Cederlane SALMONELLA O GROUP 150TESTS
SDP
Catalog No. NC1602624
Encompass
-(Product: O:57),(Reacts with factor:57),(No. of tests: 150),(SSI® SALMONELLA O GROUP ANTISERAFOR SLIDE AGGLUTINATION)
Catalog No. NC1602624 Supplier Cederlane Supplier No. 40255(SS)
-(Product: O:57),(Reacts with factor:57),(No. of tests: 150),(SSI® SALMONELLA O GROUP ANTISERAFOR SLIDE AGGLUTINATION)