Learn More
Cederlane SALMONELLA O3 COMPLEX
SDP
Catalog No. NC0486848
Encompass
(Product: O3 Complex),(Reacts with factor:3, 10, 15, 19, 34),(No. of tests: 150),(SSI� SALMONELLA O GROUP POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0486848 Supplier Cederlane Supplier No. 40219(SS)
(Product: O3 Complex),(Reacts with factor:3, 10, 15, 19, 34),(No. of tests: 150),(SSI� SALMONELLA O GROUP POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial