Learn More
Bioss Salmonella paratyphi B
SDP
Catalog No. 5000065507
Encompass_Preferred
Salmonella paratyphi B Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65507 Supplier Bioss Supplier No. BSM72396M
Salmonella paratyphi B Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide