Learn More
Bioss Salmonella paratyphi C2
SDP
Catalog No. 5000066127
Encompass_Preferred
Salmonella paratyphi C2 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66127 Supplier Bioss Supplier No. BSM72398M
Salmonella paratyphi C2 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide