Learn More
MOLTOX Molecular Toxicology Inc Salmonella Shigella Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948811
Encompass_Preferred
Salmonella Shigella (SS) Agar is a differential and selective medium used for the isolation, cultivation and differentiation of gram-negative enteric bacilli from samples like feces, urine and foods. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-811 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S011
Salmonella Shigella (SS) Agar is a differential and selective medium used for the isolation, cultivation and differentiation of gram-negative enteric bacilli from samples like feces, urine and foods. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.