Learn More
Alkali Scientific Salmonella Shigella Agar, 20 mL Monoplate, 10/Pk
SDP
Catalog No. 5000662919
Encompass_Preferred
Salmonella Shigella Agar, 20 mL Monoplate, 10/Pk. This specialized agar is designed for the isolation and identification of Salmonella and Shigella species in food and clinical samples. It supports the growth of these pathogens while inhibiting other organisms, making it an essential tool in microbiology labs.
Catalog No. 50-006-62919 Supplier Alkali Scientific Supplier No. SSAP00
Salmonella Shigella Agar, 20 mL Monoplate, 10/Pk. This specialized agar is designed for the isolation and identification of Salmonella and Shigella species in food and clinical samples. It supports the growth of these pathogens while inhibiting other organisms, making it an essential tool in microbiology labs.