Learn More
Bioworld Salmonella Shigella Agar, 500 g
Catalog No. 50254460
Encompass_Preferred
Salmonella Shigella AgarHighly selective agar used for the isolation of Salmonella and some ShigellapH: 7.0Dehydrated Appearance: tan, homogeneous powderPrepared Appearance: tan-orange to tan-red, clear to moderately hazy agarSoluble in boiling waterComposition:Beef Extract Pancreatic Digest of Tissue Peptic Digest of Animal Tissue Lactose Bile Salts Mixture Sodium Citrate Sodium Thiosulfate Ferric Citrate Brilliant Green Neutral Red Agar
Catalog No. 50-254-460 Supplier Bioworld Supplier No. 306230031
Salmonella Shigella AgarHighly selective agar used for the isolation of Salmonella and some ShigellapH: 7.0Dehydrated Appearance: tan, homogeneous powderPrepared Appearance: tan-orange to tan-red, clear to moderately hazy agarSoluble in boiling waterComposition:Beef Extract Pancreatic Digest of Tissue Peptic Digest of Animal Tissue Lactose Bile Salts Mixture Sodium Citrate Sodium Thiosulfate Ferric Citrate Brilliant Green Neutral Red Agar