Learn More
Cusabio Technology LLC salmonella typhi antibody IgM
Catalog No. 5000052092
Encompass_Preferred
The Human Salmonella Typhi (ST) Antibody (IgM) ELISA Kit is designed to qualitatively detect IgM antibodies against Salmonella Typhi in human serum or plasma samples This ELISA kit utilizes an indirect method where the presence of ST-specific IgM antibodies is identified through a colorimetric reaction measured at an absorbance wavelength of 450 nm The assay requires a sample volume of 50-100 L and delivers results within 1 to 5 hours making it suitable for rapid screening in clinical or research settings By targeting IgM antibodies this kit aids in identifying recent or active infections supporting timely diagnosis and monitoring of typhoid fever Its standardized protocol ensures consistent performance with specificity optimized to minimize cross-reactivity and enhance reliability in detecting ST-specific immune responses
Catalog No. 50-000-52092 Supplier Cusabio Technology LLC Supplier No. CSBE15070H
The Human Salmonella Typhi (ST) Antibody (IgM) ELISA Kit is designed to qualitatively detect IgM antibodies against Salmonella Typhi in human serum or plasma samples This ELISA kit utilizes an indirect method where the presence of ST-specific IgM antibodies is identified through a colorimetric reaction measured at an absorbance wavelength of 450 nm The assay requires a sample volume of 50-100 L and delivers results within 1 to 5 hours making it suitable for rapid screening in clinical or research settings By targeting IgM antibodies this kit aids in identifying recent or active infections supporting timely diagnosis and monitoring of typhoid fever Its standardized protocol ensures consistent performance with specificity optimized to minimize cross-reactivity and enhance reliability in detecting ST-specific immune responses