Learn More
AdipoGen Salsalate (1 g)
SDP
Supplier:  AdipoGen AGCR13574G001
Encompass_Preferred
Chemical. CAS: 552-94-3. Formula: C14H10O5. MW: 258.2. Nonacetylated salicylate. Non-steroidal anti-inflammatory drug (NSAID). Reduces pain and inflammation caused by conditions such as rheumatoid arthritis, osteoarthritis and related rheumatic conditions. Prostaglandin synthesis inhibitor in vivo. Inactivates cyclooxygenase-1 (COX-1) and -2 (COX-2). IKKbeta/NF-kappaB inhibitor (at significantly higher concentrations than required for COX inhibition). Used to target inflammation in the treatment of insulin resistance, type 2 diabetes, or rheumatic pain. It reduces blood glucose concentrations in patients with type 2 diabetes, as well as in insulin-resistant patients without diabetes. Reduces blood glucose, triglyceride, free fatty acid and C-reactive protein concentrations, improves glucose utilization and increases circulating insulin and adiponectin concentrations in obese adults at risk for the development of type 2 diabetes as well as for patients with type 2 diabetes.
Catalog No. 50-205-3941