Learn More
Cayman Chemical (±)-Salsolidine, Purity: ≥95%, CAS: 5784-74-7, 100 mg
SDP
Catalog No. 502950527
Encompass_Preferred
An alkaloid; inhibits COMT (Ki = 190 µM)
Catalog No. 50-295-0527 Supplier Cayman Chemical Supplier No. 32953100 MG
An alkaloid; inhibits COMT (Ki = 190 µM)