Learn More
Cayman Chemical -SalsolIdIn 50mg
SDP
Catalog No. 502948991
Encompass_Preferred
An alkaloid; inhibits COMT (Ki = 190 µM)
Catalog No. 50-294-8991 Supplier Cayman Chemical Supplier No. 3295350 MG
An alkaloid; inhibits COMT (Ki = 190 µM)