Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salsolinol-1-carboxylic acid | 57256-34-5 | 1 MG
SDP

Catalog No. 5000420726
Encompass_Preferred

Salsolinol-1-carboxylic acid is an endogenous alkaloid found in the central nervous system (CNS). This off-white to light yellow solid has a purity of 99.86% and is intended for research use only, not for sale to patients.

  • Endogenous alkaloid in the central nervous system
  • Molecular formula: C11H13NO4
  • Molecular weight: 223.23
  • Purity: 99.86%
  • Appearance: Off-white to light yellow solid
  • Soluble in H2O at 50 mg/mL (requires ultrasonic)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-20726 Supplier Medchemexpress LLC Supplier No. HYN23211MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salsolinol-1-carboxylic acid is an endogenous alkaloid found in the central nervous system (CNS). This off-white to light yellow solid has a purity of 99.86% and is intended for research use only, not for sale to patients.

  • Endogenous alkaloid in the central nervous system
  • Molecular formula: C11H13NO4
  • Molecular weight: 223.23
  • Purity: 99.86%
  • Appearance: Off-white to light yellow solid
  • Soluble in H2O at 50 mg/mL (requires ultrasonic)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.