Learn More
Cayman Chemical (±)-Salsolinol (hydrochloride), Purity: ≥99%, CAS: 79923-51-6, 100 mg
SDP
Catalog No. 502971647
Encompass_Preferred
A metabolite of ethanol produced in the brain that stimulates dopamineric neurons at 0.1 μM
Catalog No. 50-297-1647 Supplier Cayman Chemical Supplier No. 15597100 MG
A metabolite of ethanol produced in the brain that stimulates dopamineric neurons at 0.1 μM