Learn More
Sino Biological Salt Active SuperNuclease 50 kU
Catalog No. 5000696190
Encompass_Preferred
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).
Catalog No. 50-006-96190 Supplier Sino Biological Supplier No. SASN50
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).