Learn More
Waters Corp Waters Corporation Salt Bridge, Ag/AgCl Reference Electrode
Catalog No. 50792658 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
A salt bridge Ag/AgCl reference electrode is a crucial component in various electrochemical measurements, providing a stable and reliable reference potential. It typically consists of a silver wire coated with silver chloride, immersed in a solution containing chloride ions. This setup ensures a constant potential, making it ideal for applications requiring accurate and reproducible electrochemical readings. The salt bridge function allows ionic contact between two half-cells while preventing their contents from mixing.
  • Provides a stable and reproducible reference potential.
  • Facilitates accurate electrochemical measurements.
  • Ensures ionic contact between solutions.
  • Designed for use in analytical and research applications.
Catalog No. 50-792-658 Supplier Waters Corp Supplier No. 700001958
A salt bridge Ag/AgCl reference electrode is a crucial component in various electrochemical measurements, providing a stable and reliable reference potential. It typically consists of a silver wire coated with silver chloride, immersed in a solution containing chloride ions. This setup ensures a constant potential, making it ideal for applications requiring accurate and reproducible electrochemical readings. The salt bridge function allows ionic contact between two half-cells while preventing their contents from mixing.
  • Provides a stable and reproducible reference potential.
  • Facilitates accurate electrochemical measurements.
  • Ensures ionic contact between solutions.
  • Designed for use in analytical and research applications.