Learn More
EUROIMMUN Salt for PBS Buffer
Catalog No. ZF11001000 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Catalog No. Quantity
ZF11001000 10 g
1 options
Catalog No. ZF11001000 Supplier EUROIMMUN Supplier No. ZF11001000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
1 envelope
These materials, used to create a PBS buffer for use on some Dynex platforms, help to control assay drift, and work to provide the best sensitivity and specificity in results
Product Features:
  • Used to create a PBS buffer that stabilizes the assay to control drift on the DSX/DS2 systems
  • Easy to use, just open the packets and add to 2 liters of reagent grade water
  • Long shelf life and small size makes the additives easy to store, use, and manage
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Salt for PBS Buffer
Quantity 10 g
Packaging Single envelope