Learn More
Thomas Scientific Salt Sabouraud Dulcitol Agar with CAM and GEN, 90mm Plates, 10/pk
SDP
Catalog No. NC1971040
Encompass
S2Media Salt Sabouraud Dulcitol Agar is utilized for the enrichment of Candida spp., including Candida auris, which is an emerging fungal pathogen. This medium is also known as Salt Sabouraud Enrichment Agar or Salt SAB Agar. This formulation meets the CDC performance specifications, as applicable.
Catalog No. NC1971040 Supplier Thomas Scientific Supplier No. CHM01P656
S2Media Salt Sabouraud Dulcitol Agar is utilized for the enrichment of Candida spp., including Candida auris, which is an emerging fungal pathogen. This medium is also known as Salt Sabouraud Enrichment Agar or Salt SAB Agar. This formulation meets the CDC performance specifications, as applicable.