Learn More
MSC Buyers Products Vibrator Kit, TGS03 Spreader Compatible, Steel, 200 lb Force
Catalog No. 502837502
Encompass
This vibrator kit is designed for use with TGS03 salt spreaders. It provides 200 lb of force to ensure a smooth and even distribution of salt by shaking the hopper to break up clumps and prevent clogs, allowing material to flow freely. The kit includes all necessary wiring and mounting hardware for quick installation.
  • Compatible with TGS03 spreaders.
  • Constructed from steel.
  • Provides 200 lb of force.
  • Shakes hopper to break up clumps and clogs for free material flow.
  • Includes all wiring and mounting hardware for fast installation.
Catalog No. 50-283-7502 Supplier MSC Supplier No. 45967551
This vibrator kit is designed for use with TGS03 salt spreaders. It provides 200 lb of force to ensure a smooth and even distribution of salt by shaking the hopper to break up clumps and prevent clogs, allowing material to flow freely. The kit includes all necessary wiring and mounting hardware for quick installation.
  • Compatible with TGS03 spreaders.
  • Constructed from steel.
  • Provides 200 lb of force.
  • Shakes hopper to break up clumps and clogs for free material flow.
  • Includes all wiring and mounting hardware for fast installation.