Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Salton Wash #1

Catalog No. MP111002203 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111002203 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP111002203 Supplier MP Biomedicals™ Supplier No. 111002203
Add to Cart
Add to Cart

Salton Wash #1

  • Salton Wash #1 is supplied with GeneClean™ for Ancient DNA Kit to clean the GLASSMILK/DNA complex.
  • GeneClean™ for ANCIENT DNA Kit
  • Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS GUANIDINE THIOCYANATE 593-84-0; WATER 7732-18-5
Content And Storage Store at Room Temperature (15-30°C).
For Use With (Application) CleangLASSMILK/DNA complex
pH 6.7 - 7.5
Product Type Kits & Assays
Quantity 60 mL
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.