Learn More
Hampton Research HR2-907-03/SaltRx 1 #03 (A3), 185 ml - 1.5 M Ammonium chloride, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501220
Encompass
HR2-907-03/SaltRx 1 #03 (A3), 185 ml - 1.5 M Ammonium chloride, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-122-0 Supplier Hampton Research Supplier No. HR290703
HR2-907-03/SaltRx 1 #03 (A3), 185 ml - 1.5 M Ammonium chloride, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.